Nickel(II)chlorid Hydrat ≥99,995% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific
Gefahr

10815.14 10815.22 10815.06
10815.14EA 140 EUR
10815.14 10815.22 10815.06
Nickel(II)chlorid Hydrat ≥99,995% (Metall-Basis), Puratronic®
Nickel(II)chlorid Hydrat

Purity: 99.995% CAS No.: 69098-15-3 Molecular Formula: Cl2Ni.xH2O Formula Weight: 129.60(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: Green Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00149808

Formel: Cl₂Ni·H₂O
Molecular Weight: 147,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149808
CAS-Nummer: 69098-15-3
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,6505

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR