Rhodium(III)chlorid Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Rhodium trichloride hydrate
M-Clarity™ Qualitätsstufe = MQ100
520772-1G
520772-5G
SIAL520772-1GEA
526
EUR
SIAL520772-1G
SIAL520772-5G
Rhodium(III)chlorid Hydrat, Sigma-Aldrich®
Rhodium(III)chlorid Hydrat
Formel:
Cl₃Rh·H₂O Molecular Weight: 227,28 g/mol |
MDL:
MFCD00149839 CAS-Nummer: 20765-98-4 EINECS: 233-165-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...