Rhodium(III)chlorid Hydrat (38 - 41% Rh)

Lieferant: abcr

45-1880 45-1880-5G 45-1880-250MG
ABCR45-1880EA 0 EUR
ABCR45-1880 STRE45-1880-5G STRE45-1880-250MG
Rhodium(III)chlorid Hydrat (38 - 41% Rh)
Rhodium(III)chlorid Hydrat
Formel: Cl₃Rh·H₂O
MDL: MFCD00149839
CAS-Nummer: 20765-98-4
EINECS: 233-165-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR