Nickelsulfat Hexahydrat ≥98,0% ACS
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Nickel(II)sulfat Hexahydrat
36336.22 36336.36 36336.A4
36336.22EA 38.5 EUR
36336.22 36336.36 36336.A4
Nickelsulfat Hexahydrat ≥98,0% ACS
Nickelsulfat Hexahydrat
Purity: 98.0% min CAS No.: 10101-97-0 Molecular Formula: NiO4S.6H2O Formula Weight: 262.86 (154.77anhy) Physical Form: Powder Density: 2.070 Appearance: Blue to bluish green Odour: Odourless pH: 4.3-4.7 Merck Reference: 14,6517 UN No.: 3288 MDL No.: MFCD00149813
Formel: NiSO₄·6H₂O
Molecular Weight: 262.85 g/mol
Schmelzpunkt: 840
Dichte: 2.070
Lagertemperatur: Raumtemperatur
MDL: MFCD00149813
CAS-Nummer: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1 III
Merck Index: 14,6517