N-Acetyl-L-cystein
Lieferant: Molekula
Synonyme:
Ac-Cys-OH, (R)-(+)-N-Acetylcystein, (R)-(+)-2-(Ethanoylamino)-3-mercaptopropansäure
11557293-500G
11557293-100G
MOLE11557293-500GEA
123
EUR
MOLE11557293-500G
MOLE11557293-100G
N-Acetyl-L-cystein
N-Acetyl-L-cystein
Formel:
C₅H₉NO₃S Molecular Weight: 163,2 g/mol Siedepunkt: 371 °C (1013 hPa) Schmelzpunkt: 106…108 °C Dichte: 1,48 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004880 CAS-Nummer: 616-91-1 EINECS: 210-498-3 Merck Index: 13,00090 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...