D(-)-Glutamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
D(-)-2-Amino-1,5-pentandisäure-5-amid
, D(-)-Glutaminsäure-5-amid
, (R)-(-)-Glutamin
, (R)-(-)-2-Amino-1,5-pentandisäure-5-amid
, (R)-(-)-Glutaminsäure-5-amid
, H-D-Gln-OH
M-Clarity™ quality level = MQ100
SIALG9003-1GEA
117
EUR
SIALG9003-1G
SIALG9003-10G
SIALG9003-5G
D(-)-Glutamin, Sigma-Aldrich®
D(-)-Glutamin
Formel:
C₅H₁₀N₂O₃ Molecular Weight: 146,15 g/mol |
MDL:
MFCD00065607 CAS-Nummer: 5959-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...