Nanomycopulitine® (Konzentrat) 20 X, steril gefiltert
Lieferant: Biowest
Synonyme: 1-Cyclopropyl- 6-Fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride monohydrate, 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride monohydrate, Nanomycopulitine
L-X16-010 L-X16-100
L-X16-010EA 62.9 EUR
L-X16-010 L-X16-100
Nanomycopulitine® (Konzentrat) 20 X, steril gefiltert
Ciprofloxacin hydrochloride monohydrate
Nanomycopulitine concentrate can be used in vitro to clean already contaminated cell cultures and to
prevent recontamination during further cultivation.
  • Stable at temperatures below –20 °C
  • Shelf life of 24 months
Bitte beachten This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.
Formel: C₁₇H₁₈FN₃O₃·HCl·H₂O
Molecular Weight: 385.82 g/mol
MDL: MFCD00242856
CAS-Nummer: 86393-32-0