D(-)-Arginin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (R)-(-)-Arginin, H-D-Arg-OH, D(-)-α-Amino-δ-guanidinovaleriansäure, (R)-(-)-α-Amino-δ-guanidinovaleriansäure

A16137.03 A16137.06
A16137.03EA 56.1 EUR
A16137.03 A16137.06
D(-)-Arginin ≥98%
D(-)-Arginin

Purity: 98% CAS No.: 157-06-2 Molecular Formula: C6H14N4O2 Formula Weight: 174.20 Melting Point: ca 235° Beilstein Reference: 1725412

Formel: C₆H₁₄N₄O₂
Molecular Weight: 174,2 g/mol
Schmelzpunkt: ∼ 235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063116
CAS-Nummer: 157-06-2
EINECS: 205-866-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR