D(-)-Norleucin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
(R)-(-)-2-Aminohexansäure, (R)-(-)-α-Aminocapronsäure, H-D-Nle-OH, (R)-(-)-Norleucin
4031116.0005
4031116.0025
BACE4031116.0005EA
0
EUR
BACE4031116.0005
BACE4031116.0025
D(-)-Norleucin
D(-)-Norleucin
Formel:
C₆H₁₃NO₂ Molecular Weight: 131.17 g/mol Schmelzpunkt: 301 |
MDL:
MFCD00008099 CAS-Nummer: 327-56-0 EINECS: 206-320-9 Merck Index: 12,06800 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...