L(+)-Norleucin 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S)-(+)-α-Aminocapronsäure, (S)-(+)-Norleucin, (S)-(+)-2-Aminohexansäure, H-Nle-OH

226040010 226040050
ACRO226040010EA 53.7 EUR
ACRO226040010 ACRO226040050
L(+)-Norleucin 99+%
L(+)-Norleucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131.17 g/mol
Siedepunkt: 246
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064423
CAS-Nummer: 327-57-1
EINECS: 206-321-4
Merck Index: 13,06738

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 4 hrs)
Specific optical rotation +22° to +24° (20°C, 589 nm) (c=2, 6 N HCl)
Titration with HClO4 ≥99.0 %
Other amino acids (TLC) Not detectable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR