L(+)-Norleucin

Lieferant: BACHEM BIOCHEMICA
Achtung

Synonyme: (S)-(+)-α-Aminocapronsäure, (S)-(+)-Norleucin, (S)-(+)-2-Aminohexansäure, H-Nle-OH

4007429.0005 4007429.0025
BACE4007429.0005EA 0 EUR
BACE4007429.0005 BACE4007429.0025
L(+)-Norleucin
L(+)-Norleucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131.17 g/mol
Siedepunkt: 246
Schmelzpunkt: 113
MDL: MFCD00064423
CAS-Nummer: 327-57-1
EINECS: 206-321-4
Merck Index: 13,06738

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR