L(+)-Cystein, weißes Pulver Reagenz für die Zellkultur
Lieferant: MP Biomedicals
Achtung
Synonyme: (R)-(+)-2-Amino-3-mercaptopropansäure, H-Cys-OH, (R)-(+)-Cystein
0219464691 0219464625 0219464680 0219464690
ICNA0219464691EA 813 EUR
ICNA0219464691 ICNA0219464625 ICNA0219464680 ICNA0219464690
L(+)-Cystein, weißes Pulver Reagenz für die Zellkultur
L-Cystein
Storage: Store at Room Temperature (15-30 °C). Store Desiccated. Store under nitrogen.
L-Cysteine is a proteinogenic amino acid incorporated into proteins as directed by the genetic code. The thiol-side chain participates in a variety of oxidation/reduction reactions within the cell. The side chain participates in the formation of β bonds that modulate the secondary and ternary structure of proteins. Cysteine is essential and limiting for the formation of glutathione, an important antioxidant, within the cell.
Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.
NMDA glutamatergic receptor agonist.
  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Soluble in water: 25 mg/mL - Clear, Colorless Solution.
Formel: HSCH₂CH(NH₂)CO₂H
Molecular Weight: 121.16 g/mol
Siedepunkt: 254
Schmelzpunkt: 200…235 °C (Zers.)
Dichte: 1.523 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064306
CAS-Nummer: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard
Optical Rotation (c=2, 1N HCl)
7 - 13° pH (5% aq soln)
4-8