(R)-(-)-2-Amino-1-butanol 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: L(-)-2-Amino-1-butanol

B25212.06 B25212.14 B25212.22
B25212.06EA 29.1 EUR
B25212.06 B25212.14 B25212.22
(R)-(-)-2-Amino-1-butanol 98%
(R)-(-)-2-Amino-1-butanol

Purity: 98% CAS No.: 5856-63-3 Molecular Formula: C4H11NO Formula Weight: 89.14 Flash Point: 89°(192°F) Refractive Index: 1.4525 Odour: Amine-like pH: 11.1 Boiling Point: 176-178° Beilstein Reference: 1718929 Optical Rotation: -10° (neat) UN No.: 2735 MDL No.: MFCD00064419

Formel: C₄H₁₁NO
Molecular Weight: 89,14 g/mol
Siedepunkt: 176…178 °C
Schmelzpunkt: –2 °C
Dichte: 0,947
Flammpunkt: 89 °C (192 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064419
CAS-Nummer: 5856-63-3
EINECS: 227-476-4
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR