(R)-(-)-2-Amino-1-propanol 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: D(-)-Alaninol, D(-)-2-Amino-1-propanol

L11030.03 L11030.06 L11030.14
L11030.03EA 41.7 EUR
L11030.03 L11030.06 L11030.14
(R)-(-)-2-Amino-1-propanol 98%
(R)-(-)-2-Amino-1-propanol

Purity: 98% CAS No.: 35320-23-1 Molecular Formula: C3H9NO Formula Weight: 75.11 Flash Point: 83°(181°F) Refractive Index: 1.4493 Boiling Point: 174-176° Beilstein Reference: 1718866 Optical Rotation: -22° (c=2 in ethanol) UN No.: 2735 MDL No.: MFCD00064413

Formel: C₃H₉NO
Molecular Weight: 75,11 g/mol
Siedepunkt: 174…176 °C
Dichte: 0,965
Flammpunkt: 83 °C (181 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064413
CAS-Nummer: 35320-23-1
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR