(S)-(+)-1-Methoxy-2-propanol 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-(+)-Propylenglycolmonomethylether

331790050 331790250
ACRO331790050EA 210 EUR
ACRO331790050 ACRO331790250
(S)-(+)-1-Methoxy-2-propanol 99%
(S)-(+)-1-Methoxy-2-propanol
Formel: C₄H₁₀O₂
Molecular Weight: 90,12 g/mol
Schmelzpunkt: –97 °C
Flammpunkt: 30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632588
CAS-Nummer: 26550-55-0
UN: 3092
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4040 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +20° to +24° (25°C, 589 nm) (c=10, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR