(S)-(+)-2-Octanol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-(+)-2-Hydroxyoctan, D(+)-2-Octanol, (S)-(+)-Hexyl-methylcarbinol, D(+)-2-Hydroxyoctan, (S)-(+)-Methyl-hexylcarbinol

L12425.03 L12425.06
L12425.03EA 38.7 EUR
L12425.03 L12425.06
(S)-(+)-2-Octanol ≥99%
(S)-(+)-2-Octanol

Purity: 99% CAS No.: 6169-06-8 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 76°(169°F) Refractive Index: 1.4260 Odour: Pleasant Boiling Point: 179-180° Beilstein Reference: 1719323 Merck Reference: 14,6752 Optical Rotation: +10° (c=1 in ethanol) MDL No.: MFCD00064283

Formel: C₈H₁₈O
Molecular Weight: 130,23 g/mol
Siedepunkt: 179…180 °C
Dichte: 0,819
Flammpunkt: 76 °C (169 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064283
CAS-Nummer: 6169-06-8
EINECS: 228-213-6
UN: 1987
ADR: 3,III
Merck Index: 14,6752

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR