(S)-(+)-2-Pentanol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(S)-(+)-Propyl methyl carbinol, (S)-(+)-Methyl propyl carbinol, (S)-(+)-2-Hydroxy pentane
L09314.03
L09314.MD
L09314.03EA
176
EUR
L09314.03
L09314.MD
(S)-(+)-2-Pentanol ≥97%
(S)-(+)-2-Pentanol
Purity: 97%
CAS No.: 26184-62-3
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 33°(91°F)
Refractive Index: 1.4060
Boiling Point: 118-119°
Beilstein Reference: 1718820
Optical Rotation: +14° (neat)
UN No.: 1105
MDL No.: MFCD00065952
Formel:
C₅H₁₂O Molecular Weight: 88.15 g/mol Siedepunkt: 118…119 °C Schmelzpunkt: –50 °C Dichte: 0.810 Flammpunkt: 33 °C (91 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065952 CAS-Nummer: 26184-62-3 UN: 1105 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...