L(+)-Isoleucin 99
Lieferant: Thermo Fisher Scientific
Synonyme:
(2S,3S)-(+)-2-Amino-3-methylpentansäure, (2S,3S)-(+)-α-Amino-β-methylvaleriansäure, (2S,3S)-(+)-Isoleucin
A13699.09
A13699.18
A13699.22
A13699.30
A13699.09EA
31.4
EUR
A13699.09
A13699.18
A13699.22
A13699.30
L(+)-Isoleucin 99
L(+)-Isoleucin
Purity: 99%
CAS No.: 73-32-5
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Melting Point: ca 287° dec.
Beilstein Reference: 1721792
Merck Reference: 14,5179
Optical Rotation: +40° (c=5 in 5N HCl)
MDL No.: MFCD00064222
Formel:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Molecular Weight: 131.17 g/mol Schmelzpunkt: ∼ 287 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064222 CAS-Nummer: 73-32-5 EINECS: 200-798-2 Merck Index: 14,5179 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...