L(+)-Isoleucin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2S,3S)-(+)-2-Amino-3-methylpentansäure, (2S,3S)-(+)-α-Amino-β-methylvaleriansäure, (2S,3S)-(+)-Isoleucin
M-Clarity™ Qualitätsstufe = MQ200
I2752-100G
I2752-10G
I2752-1G
I2752-1KG
I2752-25G
I2752-5G
SIALI2752-100GEA
277
EUR
SIALI2752-100G
SIALI2752-10G
SIALI2752-1G
SIALI2752-1KG
SIALI2752-25G
SIALI2752-5G
L(+)-Isoleucin, Sigma-Aldrich®
L(+)-Isoleucin
Formel:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Molecular Weight: 131.17 g/mol Schmelzpunkt: 279…280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064222 CAS-Nummer: 73-32-5 EINECS: 200-798-2 Merck Index: 14,05179 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...