D(-)-Valin 98+%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-(-)-Valin, (R)-(-)-2-Amino-3-methylbutansäure, (R)-(-)-2-Aminoisovaleriansäure, H-D-Val-OH

227220250 227220050
ACRO227220250EA 112 EUR
ACRO227220250 ACRO227220050
D(-)-Valin 98+%
D(-)-Valin
Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Siedepunkt: 217
Schmelzpunkt: 302…303 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064219
CAS-Nummer: 640-68-6
EINECS: 211-368-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Loss on drying ≤0.5 % (105°C, 4 hrs)
Sulfated ash ≤0.2 %
TLC ≤1 % L-enantiomer
Specific optical rotation -26° to -29° (20°C, 589 nm) (c=5, 5 N HCl)
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR