D(-)-Valin ≥99%, Pulver

Lieferant: MP Biomedicals

Synonyme: (R)-(-)-Valin, (R)-(-)-2-Amino-3-methylbutansäure, (R)-(-)-2-Aminoisovaleriansäure, H-D-Val-OH

0210322625 0210322605
ICNA0210322625EA 132 EUR
ICNA0210322625 ICNA0210322605
D(-)-Valin ≥99%, Pulver
D(-)-Valin
Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Siedepunkt: 217
Schmelzpunkt: 86
MDL: MFCD00064219
CAS-Nummer: 640-68-6
EINECS: 211-368-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR