D(-)-Leucin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (R)-(-)-α-Amino-γ-methylvaleriansäure, H-D-Leu-OH, (R)-(-)-Leucin, (R)-(-)-2-Amino-4-methylpentansäure

4007218.0005 4007218.0100
BACE4007218.0005EA 0 EUR
BACE4007218.0005 BACE4007218.0100
D(-)-Leucin
D(-)-Leucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131.17 g/mol
Siedepunkt: 226
MDL: MFCD00063088
CAS-Nummer: 328-38-1
EINECS: 206-327-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR