D-Norvalin ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-(-)-2-Aminopentansäure, (R)-(-)-Norvalin, (R)-(-)-2-Aminovaleriansäure, H-D-Nva-OH

B23444.03 B23444.06 B23444.14
B23444.03EA 25.8 EUR
B23444.03 B23444.06 B23444.14
D-Norvalin ≥99%
D-Norvalin

Purity: 99% CAS No.: 2013-12-9 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Melting Point: 300° Beilstein Reference: 1721161 Merck Reference: 14,6716 Optical Rotation: -25° (c=10 in 20% HCl) MDL No.: MFCD00008097

Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008097
CAS-Nummer: 2013-12-9
EINECS: 217-936-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR