L(+)-Valin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (S)-(+)-Valin, (S)-(+)-2-Amino-3-methylbutansäure, (S)-(+)-2-Aminoisovaleriansäure, H-Val-OH

4031328.0100 4031328.0250
BACE4031328.0100EA 0 EUR
BACE4031328.0100 BACE4031328.0250
L(+)-Valin
L(+)-Valin
Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Siedepunkt: 217
Schmelzpunkt: 86
Dichte: 1.284 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064220
CAS-Nummer: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR