L(+)-Leucin ≥99%
Lieferant: Thermo Scientific
Synonyme: H-Leu-OH, (S)-(+)-α-Amino-γ-methylvaleriansäure, L-Leucin, (S)-(+)-Leucin, (S)-(+)-2-Amino-4-methylpentansäure
A12311.0E A12311.22 A12311.36
A12311.0EEA 758 EUR
A12311.0E A12311.22 A12311.36
L(+)-Leucin ≥99%
L(+)-Leucin
Purity: 99% CAS No.: 61-90-5 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Physical Form: Cryst powder Density: 1.293 Appearance: White Melting Point: 300° Beilstein Reference: 1721722 Merck Reference: 14,5451 Optical Rotation: +15° (c=2 in 5N HCl) MDL No.: MFCD00002617
Formel: C₆H₁₃NO₂
Molecular Weight: 131,17 g/mol
Schmelzpunkt: >300 °C
Dichte: 1,293
Lagertemperatur: Raumtemperatur
MDL: MFCD00002617
CAS-Nummer: 61-90-5
EINECS: 200-522-0
Merck Index: 14,5451