L(+)-Leucin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
H-Leu-OH, (S)-(+)-α-Amino-γ-methylvaleriansäure, L-Leucin, (S)-(+)-Leucin, (S)-(+)-2-Amino-4-methylpentansäure
4030792.0250
4030792.1000
BACE4030792.0250EA
0
EUR
BACE4030792.0250
BACE4030792.1000
L(+)-Leucin
L(+)-Leucin
Formel:
C₆H₁₃NO₂ Molecular Weight: 131.17 g/mol Schmelzpunkt: 300 Dichte: 1.165 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002617 CAS-Nummer: 61-90-5 EINECS: 200-522-0 Merck Index: 14,05451 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...