L-Norvalin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (S)-(+)-Norvalin, (S)-(+)-2-Aminovaleriansäure, (S)-(+)-2-Aminopentansäure, H-Nva-OH

4031120.0005 4031120.0025
BACE4031120.0005EA 0 EUR
BACE4031120.0005 BACE4031120.0025
L-Norvalin
L-Norvalin
Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Siedepunkt: 228
Schmelzpunkt: 101
MDL: MFCD00064421
CAS-Nummer: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR