D(-)-Leucinol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(R)-(-)-2-Amino-4-methyl-1-pentanol
413120010
413120050
ACRO413120010EA
37.8
EUR
ACRO413120010
ACRO413120050
D(-)-Leucinol 97%
D(-)-Leucinol
Formel:
C₆H₁₅NO Molecular Weight: 117,19 g/mol Siedepunkt: 199 °C (760 mmHg) Dichte: 0,907 g/cm³ Flammpunkt: 90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004734 CAS-Nummer: 53448-09-2 |
Spezifikation Testergebnisse
Appearance | Clear light yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4490 to 1.4530 (20°C, 589 nm) |
GC | ≥96.0 % |
Specific optical rotation | -3° to -5° (20°C, 589 nm) (c=9, ethanol) |
Titration with HClO4 | 96.0 to 104.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...