D(-)-Leucinol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(R)-(-)-2-Amino-4-methyl-1-pentanol
L13243.03
L13243.06
L13243.03EA
37.8
EUR
L13243.03
L13243.06
D(-)-Leucinol ≥97%
D(-)-Leucinol
Purity: 97%
CAS No.: 53448-09-2
Molecular Formula: C6H15NO
Formula Weight: 117.19
Flash Point: 90°(194°F)
Refractive Index: 1.4496
Boiling Point: 198-200°/690mm
Beilstein Reference: 1719241
Optical Rotation: -3.2° (c=9 in ethanol)
MDL No.: MFCD00004734
Formel:
C₆H₁₅NO Molecular Weight: 117,19 g/mol Siedepunkt: 198…200 °C (690 mmHg) Dichte: 0,917 Flammpunkt: 90 °C (194 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004734 CAS-Nummer: 53448-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...