L(+)-Histidin

Lieferant: Serva

Synonyme: (S)-(+)-2-Amino-3-(4-imidazoyl)propansäure, H, (S)-(+)-Histidin, His, (2S)-(+)-Histidin, H-His-OH, L-3-Imidazol-4-ylalanin

24820.02 24820.03
SERA24820.02EA 71.6 EUR
SERA24820.02 SERA24820.03
L(+)-Histidin
L(+)-Histidin
Formel: C₆H₉N₃O₂
Molecular Weight: 155.16 g/mol
Schmelzpunkt: 272…273 °C
Dichte: 1.44 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064315
CAS-Nummer: 71-00-1
EINECS: 200-745-3
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR