D(+)-Tryptophan
Lieferant: BACHEM BIOCHEMICA
Synonyme:
H-D-Trp-OH
4031216.0025
4031216.0100
BACE4031216.0025EA
0
EUR
BACE4031216.0025
BACE4031216.0100
D(+)-Tryptophan
D(+)-Tryptophan
Formel:
C₁₁H₁₂N₂O₂ Molecular Weight: 204.23 g/mol Schmelzpunkt: 280…283 °C |
MDL:
MFCD00005647 CAS-Nummer: 153-94-6 EINECS: 205-819-9 Merck Index: 14,09797 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...