D(+)-Tryptophan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

T9753-100G T9753-25G T9753-5G
SIALT9753-100GEA 756 EUR
SIALT9753-100G SIALT9753-25G SIALT9753-5G
D(+)-Tryptophan, Sigma-Aldrich®
D(+)-Tryptophan
Formel: C₁₁H₁₂N₂O₂
Schmelzpunkt: 280…283 °C
MDL: MFCD00005647
CAS-Nummer: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR