L(-)-Tryptophan, Millipore®
Lieferant: Merck
Synonyme:
L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
M-Clarity™ Qualitätsstufe = MQ100
1.08374.0010
1.08374.0100
1.08374.0500
1.08374.9010
1.08374.0010EA
50.8
EUR
1.08374.0010
1.08374.0100
1.08374.0500
1.08374.9010
L(-)-Tryptophan, Millipore®
L(-)-Tryptophan
Formel:
C₁₁H₁₂N₂O₂ Molecular Weight: 204,23 g/mol Schmelzpunkt: 290 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064340 CAS-Nummer: 73-22-3 EINECS: 200-795-6 Merck Index: 12,09929 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...