L(-)-Tryptophan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
M-Clarity™ Qualitätsstufe = MQ200
T8941-100G
T8941-10MG
T8941-1KG
T8941-25G
SIALT8941-100GEA
491
EUR
SIALT8941-100G
SIALT8941-10MG
SIALT8941-1KG
SIALT8941-25G
L(-)-Tryptophan, Sigma-Aldrich®
L(-)-Tryptophan
Formel:
C₁₁H₁₂N₂O₂ Molecular Weight: 204,23 g/mol Schmelzpunkt: 290 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064340 CAS-Nummer: 73-22-3 EINECS: 200-795-6 Merck Index: 12,09929 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...