(R)-(-)-2-Phenylbuttersäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(R)-(-)-2-Phenylbutanoic acid
L09464.03
L09464.06
L09464.03EA
102
EUR
L09464.03
L09464.06
(R)-(-)-2-Phenylbuttersäure ≥99%
(R)-(-)-2-Phenylbuttersäure
Purity: 99%
CAS No.: 938-79-4
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point:
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Siedepunkt: 270…272 °C Dichte: 1,055 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063165 CAS-Nummer: 938-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...