D(+)-Tyrosin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-(+)-p-Hydroxyphenylalanin, H-D-Tyr-OH, (R)-(+)-2-Amino-3-(4-hydroxyphenyl)propionsäure, D(+)-p-Hydroxyphenylalanin

226910025 226910100
ACRO226910025EA 52.9 EUR
ACRO226910025 ACRO226910100
D(+)-Tyrosin 98%
D(+)-Tyrosin
Formel: C₉H₁₁NO₃
Molecular Weight: 181.19 g/mol
Siedepunkt: 351
Schmelzpunkt: 310…314 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063073
CAS-Nummer: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 h)
Solubility (5 % in 1 M HCl) clear colorless to almost colorless
Residue after ignition ≤0.1 %
Specific optical rotation +10° to +12° (20°C, 589 nm) (c=5, 1 M HCl, after 3 h)
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR