D(+)-Tyrosin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-(+)-p-Hydroxyphenylalanin, H-D-Tyr-OH, (R)-(+)-2-Amino-3-(4-hydroxyphenyl)propionsäure, D(+)-p-Hydroxyphenylalanin

A17709.03 A17709.06 A17709.14
A17709.03EA 29.2 EUR
A17709.03 A17709.06 A17709.14
D(+)-Tyrosin ≥99%
D(+)-Tyrosin

Purity: 99% CAS No.: 556-02-5 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: 310-314° Beilstein Reference: 2212157 Merck Reference: 14,9839 Optical Rotation: +11° (c=4 in 1N HCl) MDL No.: MFCD00063073

Formel: C₉H₁₁NO₃
Molecular Weight: 181.19 g/mol
Siedepunkt: 351
Schmelzpunkt: 310…314 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063073
CAS-Nummer: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR