L-Tyrosin, EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®
Lieferant: Merck
Synonyme:
Tyr, (S)-(-)-2-Amino-3-(4-hydroxyphenyl)propionsäure, L(-)-p-Hydroxyphenylalanin, (S)-(-)-p-Hydroxyphenylalanin, Y, H-Tyr-OH
1.08378.1000
1.08378.9010
1.08378.1000EA
1060
EUR
1.08378.1000
1.08378.9010
L-Tyrosin, EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®
L(-)-Tyrosin
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol Siedepunkt: 351 °C (1013 hPa) Schmelzpunkt: 285 °C Dichte: 1,41 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002606 CAS-Nummer: 60-18-4 EINECS: 200-460-4 Merck Index: 13,09907 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...