(R)-(-)-4-Penten-2-ol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(-)-1-Penten-4-ol, (R)-(-)-Allyl-methylcarbinol
M-Clarity™ Qualitätsstufe = MQ100
558036-1G
558036-5G
SIAL558036-1GEA
146
EUR
SIAL558036-1G
SIAL558036-5G
(R)-(-)-4-Penten-2-ol, Sigma-Aldrich®
(R)-(-)-4-Penten-2-ol
Formel:
C₅H₁₀O Molecular Weight: 86,13 g/mol |
MDL:
MFCD03701536 CAS-Nummer: 64584-92-5 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...