(S)-(+)-4-Penten-2-ol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-(+)-1-Penten-4-ol, (S)-(+)-Allyl-methylcarbinol

B25728.03 B25728.06
B25728.03EA 114 EUR
B25728.03 B25728.06
(S)-(+)-4-Penten-2-ol ≥97%
(S)-(+)-4-Penten-2-ol

Purity: 97% CAS No.: 55563-79-6 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 30°(86°F) Refractive Index: 1.4250 Boiling Point: 115-116° Optical Rotation: +5.2° (c=1 in chloroform) UN No.: 1987 MDL No.: MFCD03427200

Formel: C₅H₁₀O
Molecular Weight: 86,13 g/mol
Siedepunkt: 115…116 °C
Dichte: 0,837
Flammpunkt: 30 °C (86 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03427200
CAS-Nummer: 55563-79-6
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR