(R)-(+)-3-Butin-2-ol ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L18880.03
L18880.06
L18880.MD
L18880.03EA
129
EUR
L18880.03
L18880.06
L18880.MD
(R)-(+)-3-Butin-2-ol ≥99%
(R)-(+)-3-Butin-2-ol
Purity: 99%
CAS No.: 42969-65-3
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: 28°(82°F)
Refractive Index: 1.4260
Boiling Point: 108-109°
Beilstein Reference: 3587272
Optical Rotation: +47° (neat)
UN No.: 2929
MDL No.: MFCD00211237
Formel:
C₄H₆O Siedepunkt: 108…109 °C Dichte: 0,891 Flammpunkt: 28 °C (82 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00211237 CAS-Nummer: 42969-65-3 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...