(S)-(-)-3-Butin-2-ol 95%, 98% ee
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-(-)-3-Hydroxy-1-butin, (S)-(-)-But-3-in-2-ol
362410010
362410050
ACRO362410010EA
78.5
EUR
ACRO362410010
ACRO362410050
(S)-(-)-3-Butin-2-ol 95%, 98% ee
(S)-(-)-3-Butin-2-ol
Formel:
C₄H₆O Molecular Weight: 70,09 g/mol Flammpunkt: 26 °C |
MDL:
MFCD00190166 CAS-Nummer: 2914-69-4 UN: 2929 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Enantiomeric excess | ≥97.5 % (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...