(S)-(-)-3-Butin-2-ol 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-(-)-3-Hydroxy-1-butin, (S)-(-)-But-3-in-2-ol
L20345.03
L20345.06
L20345.03EA
96.9
EUR
L20345.03
L20345.06
(S)-(-)-3-Butin-2-ol 99%
(S)-(-)-3-Butin-2-ol
Purity: 99%
CAS No.: 2914-69-4
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: 28°(82°F)
Refractive Index: 1.4260
Boiling Point: 108-109°
Beilstein Reference: 4652597
Optical Rotation: -47° (neat)
UN No.: 2929
MDL No.: MFCD00190166
Formel:
C₄H₆O Molecular Weight: 70,09 g/mol Siedepunkt: 108…109 °C Dichte: 0,891 Flammpunkt: 28 °C (82 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190166 CAS-Nummer: 2914-69-4 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...