(R)-(+)-α-Methylbenzylamin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(R)-(+)-1-Amino-1-phenylethan, (R)-(+)-1-Phenylethylamin, (R)-(+)-α-Phenylethylamin, (R)-(+)-1-Amino-1-phenylethane
A10967.14
A10967.22
A10967.36
A10967.14EA
43.8
EUR
A10967.14
A10967.22
A10967.36
(R)-(+)-α-Methylbenzylamin 98%
(R)-(+)-α-Methylbenzylamin
Purity: 98%
CAS No.: 3886-69-9
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 71°(159°F)
Refractive Index: 1.5260
Odour: Amine-like
Melting Point: -10°
Boiling Point: 187°
Beilstein Reference: 2410916
Merck Reference: 14,6026
Optical Rotation: +29° (c=10 in ethanol)
UN No.: 2735
MDL No.: MFCD00064405
Formel:
C₈H₁₁N Molecular Weight: 121,18 g/mol Siedepunkt: 187 °C Schmelzpunkt: –10 °C Dichte: 0,948 Flammpunkt: 71 °C (159 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064405 CAS-Nummer: 3886-69-9 EINECS: 223-423-4 UN: 2922 ADR: 8,II Merck Index: 14,6026 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...