(S)-(-)-α-Methylbenzylamin 99+%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-(-)-1-Phenylethylamin, (S)-(-)-1-Amino-1-phenylethane, (S)-(-)-1-Amino-1-phenylethan, (S)-(-)-α-Phenylethylamin
371452500
371450010
371450500
ACRO371452500EA
233
EUR
ACRO371452500
ACRO371450010
ACRO371450500
(S)-(-)-α-Methylbenzylamin 99+%
(S)-(-)-α-Methylbenzylamin
produced by BASF AG
Formel:
C₈H₁₁N Molecular Weight: 121,18 g/mol Siedepunkt: 187…189 °C (760 mmHg) Schmelzpunkt: –10 °C Dichte: 0,94 g/cm³ Flammpunkt: 71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064406 CAS-Nummer: 2627-86-3 EINECS: 220-098-0 UN: 2735 ADR: 8,II Merck Index: 12,06106 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5250 to 1.5270 (20°C, 589 nm) |
Water | ≤0.5 % |
GC | ≥99.0 % |
Specific optical rotation | -39° to -41° (20°C, 589 nm) (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...