(S)-(-)-α-Methylbenzylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (S)-(-)-1-Phenylethylamin, (S)-(-)-1-Amino-1-phenylethane, (S)-(-)-1-Amino-1-phenylethan, (S)-(-)-α-Phenylethylamin

A12632.14 A12632.22 A12632.36
A12632.14EA 40.9 EUR
A12632.14 A12632.22 A12632.36
(S)-(-)-α-Methylbenzylamin 98%
(S)-(-)-α-Methylbenzylamin

Purity: 98% CAS No.: 2627-86-3 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 71°(159°F) Refractive Index: 1.5260 Odour: Amine-like Melting Point: -10° Boiling Point: 187° Beilstein Reference: 2204907 Merck Reference: 14,6026 Optical Rotation: -30° (c=10 in ethanol) UN No.: 2735 MDL No.: MFCD00064406

Formel: C₈H₁₁N
Molecular Weight: 121.18 g/mol
Siedepunkt: 187 °C
Schmelzpunkt: –10 °C
Dichte: 0.948
Flammpunkt: 71 °C (159 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064406
CAS-Nummer: 2627-86-3
EINECS: 220-098-0
UN: 2922
ADR: 8 II
Merck Index: 12,06106

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR