(S)-(-)-1-Phenylethanol ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (S)-(-)-sec-Phenethylalkohol, (S)-(-)-α-Methylbenzylalkohol

B21188.03 B21188.06 B21188.14
B21188.03EA 31 EUR
B21188.03 B21188.06 B21188.14
(S)-(-)-1-Phenylethanol ≥98%
(S)-(-)-1-Phenylethanol

Purity: 99% CAS No.: 1445-91-6 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 85°(185°F) Refractive Index: 1.5280 Odour: Mild Melting Point: 9-11° Boiling Point: 97-99°/20mm Beilstein Reference: 2039797 Optical Rotation: -45° (c=5 in methanol) UN No.: 2937 MDL No.: MFCD00064264

Formel: C₈H₁₀O
Molecular Weight: 122,17 g/mol
Siedepunkt: 97…99 °C (20 mmHg)
Schmelzpunkt: 9…11 °C
Dichte: 1,012
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064264
CAS-Nummer: 1445-91-6
UN: 2937
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR