(R)-(-)-2-Methylpiperazin
Lieferant: FLUOROCHEM
Synonyme:
(R)-2-Methylpiperazine
011823-10G
011823-1G
FLUO011823-10GEA
67.1
EUR
FLUO011823-10G
FLUO011823-1G
(R)-(-)-2-Methylpiperazin
(R)-(-)-2-Methylpiperazin
Formel:
C₅H₁₂N₂ Molecular Weight: 100,16 g/mol Siedepunkt: 153 °C (1013 hPa) Schmelzpunkt: 89…91 °C |
MDL:
MFCD00192317 CAS-Nummer: 75336-86-6 UN: 1325 ADR: 4.1,II |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...