1,10-Phenanthrolin, (max. 1,5% H₂O) 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: o-Phenanthroline

A13163.22 A13163.14 A13163.06
A13163.22EA 314 EUR
A13163.22 A13163.14 A13163.06
1,10-Phenanthrolin, (max. 1,5% H₂O) 99%
1,10-Phenanthrolin

1,10-Phenanthroline, anhydrous is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.


Purity: 99% CAS No.: 66-71-7 Molecular Formula: C12H8N2 Formula Weight: 180.21 Odour: Odourless Melting Point: 116-119° Boiling Point: 330° Beilstein Reference: 126461 Merck Reference: 14,7214 UN No.: 2811 MDL No.: MFCD00011678

Formel: C₁₂H₈N₂
Molecular Weight: 180,21 g/mol
Siedepunkt: >330 °C
Schmelzpunkt: 116…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011678
CAS-Nummer: 66-71-7
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,7214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5092a8ec81301ed6ed30f75429f9f27784db47a5&vid=106c4325bb7c310de902d3118cf4b31bcee3dc92&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR