4-Methyl-1,10-phenanthrolin
Lieferant: Thermo Scientific
|
Achtung
|
31888.03
31888.06
31888.03EA
80.3
EUR
31888.03
31888.06
4-Methyl-1,10-phenanthrolin
4-Methyl-1,10-phenanthroline
CAS No.: 31301-28-7
Molecular Formula: C13H10N2
Formula Weight: 194.24
Physical Form: Crystalline
Melting Point: 144-145°
MDL No.: MFCD00004978
Formel:
C₁₃H₁₀N₂ Schmelzpunkt: 144…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004978 CAS-Nummer: 31301-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...